Order Entry
Export
Orders LinkContactUsLinkComponent
MES sodium salt

MES sodium salt

Glass bottle for solids, 100 g
Catalog # ROTH6922.2
Supplier:   Roth Carl
MES sodium salt
Catalog # ROTH6922.2
Supplier:  Roth Carl
CAS Number:  71119-23-8
MES sodium salt

Glass bottle for solids, 100 g

CAS Number:  71119-23-8
Price
109,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
  • CAS Number:71119-23-8
  • Formula:C₆H₁₂NNaO₄S
  • MDL Number:MFCD00065473
  • Molecular Weight:217.22 g/mol
  • EINECS:275-203-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications