MES sodium salt
Glass bottle for solids, 25 g
Catalog # ROTH6922.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
- CAS Number:71119-23-8
- Formula:C₆H₁₂NNaO₄S
- MDL Number:MFCD00065473
- Molecular Weight:217.22 g/mol
- EINECS:275-203-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



