Order Entry
Export
Orders LinkContactUsLinkComponent
Lactose monohydrate

Lactose monohydrate

5 kg
Catalog # ROTH6868.2
Supplier:   Roth Carl
Lactose monohydrate
Catalog # ROTH6868.2
Supplier:  Roth Carl
CAS Number:  10039-26-6
Lactose monohydrate

5 kg

CAS Number:  10039-26-6
Price
117,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Milk sugar, Lactobiose
  • CAS Number:10039-26-6
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • Molecular Weight:360.32 g/mol
  • Melting Pt:223 °C
  • Storage Temperature:Ambient
  • EINECS:200-559-2
Specifications

Specifications

  • Pk:
    5 kg

Specifications