Lactose monohydrate
Glass bottle for solids, 1 kg
Catalog # ROTH6868.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Milk sugar, Lactobiose
- CAS Number:10039-26-6
- Formula:C₁₂H₂₂O₁₁·1H₂O
- Molecular Weight:360.32 g/mol
- Melting Pt:223 °C
- Storage Temperature:Ambient
- EINECS:200-559-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



