Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl acetate

Ethyl acetate

5 L
Catalog # ROTH6784.5
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   Roth Carl
Ethyl acetate
Catalog # ROTH6784.5
Supplier:  Roth Carl
   Safer Human and Environmental Health
CAS Number:  141-78-6
Ethyl acetate
Ethyl acetate

5 L

CAS Number:  141-78-6
Price
128,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications

Specifications

  • Pk:
    5 L
  • Cat. No.:
    ROTH6784.5
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications