Potassium chloride
Glass bottle for solids, 500 g
ROTH6781.3
: Potassiumchloride
- :7447-40-7
- :KCl
- :MFCD00011360
- :74.55 g/mol
- :14,07621
- :1420 °C (1013 hPa)
- :773 °C
- :1.98 g/cm³ (20 °C)
- :Ambient
- :231-211-8
- Pack type:Glass bottle for solids
- Pk:500 g