Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium chloride

Potassium chloride

Glass bottle for solids, 1 kg
Catalog # ROTH6781.1
Supplier:   Roth Carl
Potassium chloride
Catalog # ROTH6781.1
Supplier:  Roth Carl
CAS Number:  7447-40-7
Potassium chloride

Glass bottle for solids, 1 kg

CAS Number:  7447-40-7
Price
38,30 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassiumchloride
  • CAS Number:7447-40-7
  • Formula:KCl
  • MDL Number:MFCD00011360
  • Molecular Weight:74.55 g/mol
  • Merck Index:14,07621
  • Boiling Pt:1420 °C (1013 hPa)
  • Melting Pt:773 °C
  • Density:1.98 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-211-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications