Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium iodide

Potassium iodide

Glass bottle for solids, 1 kg
Catalog # ROTH6750.2
Supplier:   Roth Carl
Potassium iodide
Catalog # ROTH6750.2
Supplier:  Roth Carl
CAS Number:  7681-11-0
Potassium iodide

Glass bottle for solids, 1 kg

CAS Number:  7681-11-0
Price
390,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:686 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications