Adenosine
Glass bottle for solids, 25 g
ROTH6122.3
: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- :58-61-7
- :C₁₀H₁₃N₅O₄
- :MFCD00005752
- :267.24 g/mol
- :Refrigerator
- :200-389-9
- Pack type:Glass bottle for solids
- Pk:25 g