Order Entry
Export
Orders LinkContactUsLinkComponent
Adenosine

Adenosine

Glass bottle for solids, 5 g
Catalog # ROTH6122.1
Supplier:   Roth Carl
Adenosine
Catalog # ROTH6122.1
Supplier:  Roth Carl
CAS Number:  58-61-7
Adenosine

Glass bottle for solids, 5 g

CAS Number:  58-61-7
Price
22,40 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-389-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications