Palmitic acid
5 kg
Catalog # ROTH5907.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Hexadecanoic acid, Hexadecylic acid
- CAS Number:57-10-3
- Formula:C₁₆H₃₂O₂
- MDL Number:MFCD00002747
- Molecular Weight:256.43 g/mol
- Merck Index:13,07064
- Boiling Pt:352 °C (1013 hPa)
- Melting Pt:61…63 °C
- Density:0.853 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-312-9
Specifications
Specifications
- Pk:5 kg



