Order Entry
Export
Orders LinkContactUsLinkComponent
Benzene

Benzene

Glass bottle, 100 mL
Catalog # ROTH5785.3
Flammable
Health
Supplier:   Roth Carl
Benzene
Catalog # ROTH5785.3
Supplier:  Roth Carl
CAS Number:  71-43-2
Benzene
Benzene

Glass bottle, 100 mL

CAS Number:  71-43-2
Price
26,50 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Cyclohexatriene
  • CAS Number:71-43-2
  • Formula:C₆H₆
  • MDL Number:MFCD00003009
  • Molecular Weight:78.11 g/mol
  • Merck Index:13,01066
  • Boiling Pt:80.1 °C (1013 hPa)
  • Melting Pt:5.5 °C
  • Density:0.878 g/cm³ (20 °C)
  • Flash Pt:–11 °C
  • Storage Temperature:Ambient
  • EINECS:200-753-7
  • UN:1114
  • ADR:3,II
  • REACH:01-2119447106-44
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications