Order Entry
Export
Orders LinkContactUsLinkComponent
Benzoic acid

Benzoic acid

Glass bottle for solids, 1 kg
Catalog # ROTH5781.1
Irritant
Supplier:   Roth Carl
Benzoic acid
Catalog # ROTH5781.1
Supplier:  Roth Carl
CAS Number:  65-85-0
Benzoic acid
Benzoic acid

Glass bottle for solids, 1 kg

CAS Number:  65-85-0
Price
55,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:65-85-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00002398
  • Molecular Weight:122.12 g/mol
  • Merck Index:13,01092
  • Boiling Pt:249 °C (1013 hPa)
  • Melting Pt:121…123 °C
  • Density:1.316 g/cm³ (20 °C)
  • Flash Pt:121 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-618-2
  • REACH:01-2119455536-33
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications