Order Entry
Export
Orders LinkContactUsLinkComponent
Bacitracin

Bacitracin

Glass bottle for solids, 25 g
Catalog # ROTH5655.2
Supplier:   Roth Carl
Bacitracin
Catalog # ROTH5655.2
Supplier:  Roth Carl
CAS Number:  1405-87-4
Bacitracin

Glass bottle for solids, 25 g

CAS Number:  1405-87-4
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:1405-87-4
  • Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL Number:MFCD00062640
  • Molecular Weight:1421.60 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications