Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxycoumarin

7-Hydroxycoumarin

Glass bottle for solids, 100 g
Catalog # ROTH5437.2
Supplier:   Roth Carl
7-Hydroxycoumarin
Catalog # ROTH5437.2
Supplier:  Roth Carl
CAS Number:  93-35-6
7-Hydroxycoumarin

Glass bottle for solids, 100 g

CAS Number:  93-35-6
Price
342,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162.14 g/mol
  • Merck Index:12,09981
  • Boiling Pt:382 °C (1013 hPa)
  • Melting Pt:204…206 °C
  • Storage Temperature:Ambient
  • EINECS:202-240-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications