Glutaraldehyde
Glass bottle, 2,5 L
Catalog # ROTH4995.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
- CAS Number:111-30-8
- Formula:C₅H₈O₂
- MDL Number:MFCD00007025
- Molecular Weight:100.12 g/mol
- Merck Index:13,04485
- Boiling Pt:188 °C (1013 hPa)
- Melting Pt:–10 °C
- Density:1.014 g/cm³ (20 °C)
- Storage Temperature:Freezer
- EINECS:203-856-5
- ADR:6.1,II
- :Density: 1.014 g/cm³ (20 °C)
- UN:2927
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



