Order Entry
Export
Orders LinkContactUsLinkComponent
Glutaraldehyde

Glutaraldehyde

Glass bottle, 500 mL
Catalog # ROTH4995.1
Toxic
Corrosive
Health
Environment
Supplier:   Roth Carl
Glutaraldehyde
Catalog # ROTH4995.1
Supplier:  Roth Carl
CAS Number:  111-30-8
Glutaraldehyde

Glass bottle, 500 mL

CAS Number:  111-30-8
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Glutaral, Glutardialdehyde, 1,5-Pentanedial
  • CAS Number:111-30-8
  • Formula:C₅H₈O₂
  • MDL Number:MFCD00007025
  • Molecular Weight:100.12 g/mol
  • Merck Index:13,04485
  • Boiling Pt:188 °C (1013 hPa)
  • Melting Pt:–10 °C
  • Density:1.014 g/cm³ (20 °C)
  • Storage Temperature:Freezer
  • EINECS:203-856-5
  • ADR:6.1,II
  •  :Density: 1.014 g/cm³ (20 °C)
  • UN:2927
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications