n-Pentane
Glass bottle, 1 L
Catalog # ROTH4978.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Pentane
- CAS Number:109-66-0
- Formula:H₃C(CH₂)₃CH₃
- MDL Number:MFCD00009498
- Molecular Weight:72.15 g/mol
- Merck Index:13,07193
- Boiling Pt:35…36 °C (1013 hPa)
- Melting Pt:–130 °C
- Density:0.63 g/cm³ (20 °C)
- Flash Pt:–40 °C
- Storage Temperature:Ambient
- EINECS:203-692-4
- UN:1265
- ADR:3,II
- REACH:01-2119459286-30
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



