Order Entry
Export
Orders LinkContactUsLinkComponent
Barbituric acid

Barbituric acid

Glass bottle for solids, 500 g
Catalog # ROTH4927.2
Supplier:   Roth Carl
Barbituric acid
Catalog # ROTH4927.2
Supplier:  Roth Carl
CAS Number:  67-52-7
Barbituric acid

Glass bottle for solids, 500 g

CAS Number:  67-52-7
Price
394,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
  • CAS Number:67-52-7
  • Formula:C₄H₄N₂O₃
  • MDL Number:MFCD00006666
  • Molecular Weight:128.09 g/mol
  • Merck Index:13,00966
  • Boiling Pt:441 °C (1013 hPa)
  • Melting Pt:250…252 °C
  • Storage Temperature:Ambient
  • EINECS:200-658-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications