Order Entry
Export
Orders LinkContactUsLinkComponent
 

L(+)-Valine

Glass bottle for solids, 25 g
Catalog # ROTH4879.1
Supplier:   Roth Carl
L(+)-Valine
Catalog # ROTH4879.1
Supplier:  Roth Carl
CAS Number:  72-18-4
L(+)-Valine

Glass bottle for solids, 25 g

CAS Number:  72-18-4
Price
21,70 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS Number:72-18-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Boiling Pt:217 °C (1013 hPa)
  • Melting Pt:86 °C
  • Density:1.284 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-773-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications