L(-)-Tryptophan
Glass bottle for solids, 1 kg
Catalog # ROTH4858.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
- CAS Number:73-22-3
- Formula:C₁₁H₁₂N₂O₂
- MDL Number:MFCD00064340
- Molecular Weight:204.23 g/mol
- Merck Index:12,09929
- Melting Pt:290 °C
- Density:1.34 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-795-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



