Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Tryptophan

L(-)-Tryptophan

Glass bottle for solids, 10 g
Catalog # ROTH4858.1
Supplier:   Roth Carl
L(-)-Tryptophan
Catalog # ROTH4858.1
Supplier:  Roth Carl
CAS Number:  73-22-3
L(-)-Tryptophan

Glass bottle for solids, 10 g

CAS Number:  73-22-3
Price
18,60 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS Number:73-22-3
  • Formula:C₁₁H₁₂N₂O₂
  • MDL Number:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Merck Index:12,09929
  • Melting Pt:290 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-795-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications