Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Sucrose

D-(+)-Sucrose

50 kg
Catalog # ROTH4661.3
Supplier:   Roth Carl
D-(+)-Sucrose
Catalog # ROTH4661.3
Supplier:  Roth Carl
CAS Number:  57-50-1
D-(+)-Sucrose
D-(+)-Sucrose

50 kg

CAS Number:  57-50-1
Price
411,00 €
Log in for availability
1 * 50 kg
Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-334-9
Specifications

Specifications

  • Pk:
    50 kg

Specifications