1,4-Dioxane
Glass bottle, 2,5 L
Catalog # ROTH4626.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- CAS Number:123-91-1
- Formula:C₄H₈O₂
- MDL Number:MFCD00006571
- Molecular Weight:88.11 g/mol
- Merck Index:13,03330
- Boiling Pt:101.1 °C (1013 hPa)
- Melting Pt:11.8 °C
- Density:1.03 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:204-661-8
- UN:1165
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



