Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dioxane

1,4-Dioxane

Glass bottle, 1 L
Catalog # ROTH4626.1
Flammable
Health
Irritant
Supplier:   Roth Carl
1,4-Dioxane
Catalog # ROTH4626.1
Supplier:  Roth Carl
CAS Number:  123-91-1
1,4-Dioxane
1,4-Dioxane

Glass bottle, 1 L

CAS Number:  123-91-1
Price
122,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS Number:123-91-1
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03330
  • Boiling Pt:101.1 °C (1013 hPa)
  • Melting Pt:11.8 °C
  • Density:1.03 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:204-661-8
  • UN:1165
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications