Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium metaphosphate

Sodium metaphosphate

5 kg
Catalog # ROTH4356.2
Supplier:   Roth Carl
Sodium metaphosphate
Catalog # ROTH4356.2
Supplier:  Roth Carl
CAS Number:  10361-03-2
Sodium metaphosphate

5 kg

CAS Number:  10361-03-2
Price
107,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:10361-03-2
  • Formula:NaPO₃
  • Molecular Weight:101.96 g/mol
  • Melting Pt:625 °C
  • Density:2.54 g/cm³ (20 °C)
  •  :Density: 2.54 g/cm³ (20 °C)
  • EINECS:233-782-9
Specifications

Specifications

  • Pk:
    5 kg

Specifications