Sodium metaphosphate
Glass bottle for solids, 1 kg
Catalog # ROTH4356.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:10361-03-2
- Formula:NaPO₃
- Molecular Weight:101.96 g/mol
- Melting Pt:625 °C
- Density:2.54 g/cm³ (20 °C)
- :Density: 2.54 g/cm³ (20 °C)
- EINECS:233-782-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



