Gelatin
5 kg
Catalog # ROTH4308.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:5 kg
- Cat. No.:ROTH4308.2
- Protein/peptide name:Gelatin
- Storage conditions:Ambient
- Protein synonyms:Gelatine
- CAS No.:9000-70-8



