Gelatin
Glass bottle for solids, 1 kg
Catalog # ROTH4308.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Cat. No.:ROTH4308.1
- Protein/peptide name:Gelatin
- Storage conditions:Ambient
- Protein synonyms:Gelatine
- CAS No.:9000-70-8



