Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Histidine

L(+)-Histidine

Glass bottle for solids, 100 g
Catalog # ROTH3852.3
Supplier:   Roth Carl
L(+)-Histidine
Catalog # ROTH3852.3
Supplier:  Roth Carl
CAS Number:  71-00-1
L(+)-Histidine
L(+)-Histidine

Glass bottle for solids, 100 g

CAS Number:  71-00-1
Price
60,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications