Orders LinkContactUsLinkComponent
Glutaraldehyde 25% in aqueous solution
5 L
  ROTH3778.2
 :   Roth Carl
Glutaraldehyde 25% in aqueous solution
  ROTH3778.2
 :  Roth Carl
 :  111-30-8
Glutaraldehyde 25% in aqueous solution
5 L
 :  111-30-8
384,00 €
1 * 5 L
 :  Glutaral, Glutardialdehyde, 1,5-Pentanedial
  •  :111-30-8
  •  :C₅H₈O₂
  •  :MFCD00007025
  •  :100.12 g/mol
  •  :101 °C (1013 hPa)
  •  :–6 °C
  •  :1.06 g/cm³ (20 °C)
  •  :>71 °C
  •  :Refrigerator
  •  :203-856-5
  •  :6.1,II
  •  :Density: 1.014 g/cm³ (20 °C)
  •  :2927
  • Pk:
    5 L