L(+)-Glutamic acid
Glass bottle for solids, 250 g
Catalog # ROTH3774.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
- CAS Number:56-86-0
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- MDL Number:MFCD00002634
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:160 °C
- Density:1.538 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-293-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



