Acetic acid
Glass bottle, 2,5 L
Catalog # ROTH3738.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Boiling Pt:118 °C (1013 hPa)
- Melting Pt:17 °C
- Density:1.05 g/cm³ (20 °C)
- Flash Pt:38.5 °C
- Storage Temperature:Ambient
- EINECS:200-580-7
- UN:2789
- ADR:8,II
- REACH:01-2119475328-30
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



