Methyl acetate
Glass bottle, 2,5 L
Catalog # ROTH3730.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Acetic acid methyl ester
- CAS Number:79-20-9
- Formula:C₃H₆O₂
- MDL Number:MFCD00008711
- Molecular Weight:74.08 g/mol
- Merck Index:13,06038
- Boiling Pt:56.9 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.938 g/cm³ (20 °C)
- Flash Pt:–13 °C
- Storage Temperature:Ambient
- EINECS:201-185-2
- UN:1231
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



