Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl acetate

Methyl acetate

Glass bottle, 1 L
Catalog # ROTH3730.1
Flammable
Irritant
Supplier:   Roth Carl
Methyl acetate
Catalog # ROTH3730.1
Supplier:  Roth Carl
CAS Number:  79-20-9
Methyl acetate

Glass bottle, 1 L

CAS Number:  79-20-9
Price
42,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Acetic acid methyl ester
  • CAS Number:79-20-9
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00008711
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,06038
  • Boiling Pt:56.9 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.938 g/cm³ (20 °C)
  • Flash Pt:–13 °C
  • Storage Temperature:Ambient
  • EINECS:201-185-2
  • UN:1231
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications