Order Entry
Export
Orders LinkContactUsLinkComponent
Calcium hydroxide

Calcium hydroxide

Glass bottle for solids, 1 kg
Catalog # ROTH3529.4
Corrosive
Irritant
Supplier:   Roth Carl
Calcium hydroxide
Catalog # ROTH3529.4
Supplier:  Roth Carl
CAS Number:  1305-62-0
Calcium hydroxide

Glass bottle for solids, 1 kg

CAS Number:  1305-62-0
Price
19,80 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:13,01675
  • Melting Pt:550 °C
  • Density:2.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications