Calcium hydroxide
Glass bottle for solids, 2,5 kg
Catalog # ROTH3529.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Slaked lime, Calcium dihydroxide
- CAS Number:1305-62-0
- Formula:Ca(OH)₂
- MDL Number:MFCD00010901
- Molecular Weight:74.09 g/mol
- Merck Index:13,01675
- Melting Pt:550 °C
- Density:2.24 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-137-3
- UN:3262
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 kg



