n-Decane
Glass bottle, 500 mL
Catalog # ROTH3475.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:124-18-5
- Formula:H₃C(CH₂)₈CH₃
- MDL Number:MFCD00008954
- Molecular Weight:142.28 g/mol
- Boiling Pt:174.1 °C (1013 hPa)
- Melting Pt:–30 °C
- Density:0.73 g/cm³ (20 °C)
- Flash Pt:46 °C
- Storage Temperature:Ambient
- EINECS:204-686-4
- UN:2247
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



