Order Entry
Export
Orders LinkContactUsLinkComponent
n-Decane

n-Decane

Glass bottle, 100 mL
Catalog # ROTH3475.1
Flammable
Health
Supplier:   Roth Carl
n-Decane
Catalog # ROTH3475.1
Supplier:  Roth Carl
CAS Number:  124-18-5
n-Decane
n-Decane

Glass bottle, 100 mL

CAS Number:  124-18-5
Price
34,20 €
Log in for availability
1 * 100 mL
Product Details & Documents
  • CAS Number:124-18-5
  • Formula:H₃C(CH₂)₈CH₃
  • MDL Number:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:174.1 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:0.73 g/cm³ (20 °C)
  • Flash Pt:46 °C
  • Storage Temperature:Ambient
  • EINECS:204-686-4
  • UN:2247
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications