Order Entry
Export
Orders LinkContactUsLinkComponent
DL-Malic acid

DL-Malic acid

Glass bottle for solids, 1 kg
Catalog # ROTH3034.2
Corrosive
Irritant
Supplier:   Roth Carl
DL-Malic acid
Catalog # ROTH3034.2
Supplier:  Roth Carl
CAS Number:  617-48-1
DL-Malic acid
DL-Malic acid

Glass bottle for solids, 1 kg

CAS Number:  617-48-1
Price
55,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
  • CAS Number:617-48-1
  • Formula:HOOCCH(OH)CH₂COOH
  • MDL Number:MFCD00064212
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:131…132 °C
  • Density:1.601 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:210-514-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications