Order Entry
Export
Orders LinkContactUsLinkComponent
Urea

Urea

5 kg
Catalog # ROTH2317.2
Supplier:   Roth Carl
Urea
Catalog # ROTH2317.2
Supplier:  Roth Carl
CAS Number:  57-13-6
Urea
CAS Number:  57-13-6
Price
265,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specifications

  • Pk:
    5 kg

Specifications