Order Entry
Export
Orders LinkContactUsLinkComponent
Ethidium bromide 10 mg/ml in aqueous solution

Ethidium bromide 10 mg/ml in aqueous solution

Glass bottle, 10 mL
Catalog # ROTH2218.1
Toxic
Health
Supplier:   Roth Carl
Ethidium bromide 10 mg/ml in aqueous solution
Catalog # ROTH2218.1
Supplier:  Roth Carl
CAS Number:  1239-45-8
Ethidium bromide 10 mg/ml in aqueous solution

Glass bottle, 10 mL

CAS Number:  1239-45-8
Price
14,50 €
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • EINECS:214-984-6

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL

Specifications