Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Split, with glass frit, universal for Varian 1177, 6,3 mm, 4,0 mm, 78,5 mm
  568-2357
 :   Restek
Inlet liners
  568-2357
 :  Restek
Inlet liners
Inlet liners

Split, with glass frit, universal for Varian 1177, 6,3 mm, 4,0 mm, 78,5 mm

 
268,00 €
   
1 * 5 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Split, with glass frit, universal for Varian 1177
  • Length:
    78,5 mm
  • Seq:
    063