Orders LinkContactUsLinkComponent
Liner seals
Liner seals
Graphite liner seals, for Bruker/Varian 1078/1079 GCs, 450 °C
Catalog # REST22683
Supplier:   Restek
Liner seals
Catalog # REST22683
Supplier:  Restek
Liner seals
Liner seals
Graphite liner seals, for Bruker/Varian 1078/1079 GCs, 450 °C
Price
73,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Specifications
  • Temperature Range:
    450 °C
  • Description:
    Graphite liner seals, for Bruker/Varian 1078/1079 GCs
  • Seq:
    01