Orders LinkContactUsLinkComponent
Micro pipettes, end-to-end
Micro pipettes, end-to-end
20 µl, 29 ±1 mm
Catalog # MARI2911110
Supplier:   MARIENFELD
Micro pipettes, end-to-end
Catalog # MARI2911110
Supplier:  MARIENFELD
Micro pipettes, end-to-end
Micro pipettes, end-to-end
20 µl, 29 ±1 mm
Price
131,33 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These micro pipettes made of glass can dispense more accurately than conventional pipettes. They are used for dispensing very small amounts of liquids.
  • For single use
  • Sodium-heparinized (80 iu/ml)
 : Compliance with DIN ISO 7550. Approved for in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark.
 : 100 pieces in a colour coded dispenser vial, 10 vials in a box.
Specifications
  • Capacity:
    20 µl
  • Length:
    29 ±1 mm