Septum caps, Nalgene®
38 - 430
Supplier: Thermo Fisher Scientific
Septum caps, Nalgene®
Catalog # NALG342178-0384
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
HDPE with silicone/PTFE septum.
- Sterile 10⁻⁶ SAL
- No adhesives used to attach septum to high density polyethylene closure
- Non-cytotoxic and non-pyrogenic
- Dual septum material for improved gas barrier
Specifications
- Cap Size:38 - 430
- Cat. No.:NALG342178-0384
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact