Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Double taper for trace, active samples up to 4 µl, 8,0 mm, 4,0 mm, 105 mm
  REST20952
 :   Restek
Inlet liners
  REST20952
 :  Restek
Inlet liners
Inlet liners

Double taper for trace, active samples up to 4 µl, 8,0 mm, 4,0 mm, 105 mm

 
 
   
1 * 1 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    8,0 mm
  • Ø int.:
    4,0 mm
  • Description:
    Double taper for trace, active samples up to 4 µl
  • Length:
    105 mm
  • Seq:
    108