Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL, 8,0 mm, 5,0 mm, 105 mm
  REST20947
 :   Restek
Inlet liners
  REST20947
 :  Restek
Inlet liners
Inlet liners

Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL, 8,0 mm, 5,0 mm, 105 mm

 
 
   
1 * 25 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    8,0 mm
  • Ø int.:
    5,0 mm
  • Description:
    Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL
  • Length:
    105 mm
  • Seq:
    112