Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Volumetric flask, BLAUBRAND®, A, DE-M, 50 ml, Boro 3.3, W, NS 14/23, Boro 3.3, glass stopper, ISO individual certificate 1 * 1 items
BRND937288
:
Brand
Volumetric flask, BLAUBRAND®, A, DE-M, 50 ml, Boro 3.3, W, NS 14/23, Boro 3.3, glass stopper, ISO individual certificate 1 * 1 items
BRND937288
:
Brand