Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
25 ml, 10/19, ±0,040 ml
Catalog # BRND936407
Supplier: Brand
Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
Catalog # BRND936407
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3,3 with PP or glass stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Individually certified, individual certificate included
* Wide neck
Certifications: DIN EN ISO 1042
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
Specifications
- NS:10/19
- Capacity:25 ml
- Seq:04
- Tolerance:±0,040 ml



